1-Ethoxy-4-ethyl-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

71467-500MG 71467-1G 71467-5G 71467-25G
77333-612EA 157.59 USD
77333-612 77345-720 77369-624 77390-574
1-Ethoxy-4-ethyl-2-iodobenzene ≥97%
1-Ethoxy-4-ethyl-2-iodobenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃IO
MW: 276.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764141
CAS Number: 2384877-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR