(2-Ethoxy-5-ethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

71466-500MG 71466-1G 71466-5G 71466-25G
77333-610EA 157.59 USD
77333-610 77345-670 77369-622 77390-572
(2-Ethoxy-5-ethylphenyl)(methyl)sulfane ≥97%
(2-Ethoxy-5-ethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₁H₁₆OS
Storage Temperature: Ambient
MDL Number: MFCD34764140
CAS Number: 2384731-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR