(3-Bromo-2-chloro-6-fluoro-5-methylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

71434-500MG 71434-1G 71434-5G 71434-25G
77333-608EA 197.76 USD
77333-608 77357-748 77369-620 77390-570
(3-Bromo-2-chloro-6-fluoro-5-methylphenyl)methanol ≥95%
(3-Bromo-2-chloro-6-fluoro-5-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₈H₇BrClFO
MW: 253.4959 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764132
CAS Number: 2901082-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR