2,2,2-Trifluoro-1-(4-methoxy-3-(methylthio)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

71429-500MG 71429-1G 71429-5G 71429-25G
77333-606EA 231.75 USD
77333-606 77357-746 77369-618 77390-568
2,2,2-Trifluoro-1-(4-methoxy-3-(methylthio)phenyl)ethanone ≥97%
2,2,2-Trifluoro-1-(4-methoxy-3-(methylthio)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂S
MW: 250.2374 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764130
CAS Number: 2901065-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR