3-Chloro-4-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

71413-500MG 71413-1G 71413-5G
77337-542EA 418.18 USD
77337-542 77357-744 77381-690
3-Chloro-4-iodobenzaldehyde ≥97%
3-Chloro-4-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClIO
MW: 266.464 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18393322
CAS Number: 148334-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR