2-Chloro-4-(methylthio)pyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

71411-500MG 71411-1G 71411-5G 71411-25G
77333-604EA 247.2 USD
77333-604 77357-742 77369-616 77390-566
2-Chloro-4-(methylthio)pyridin-3-ol ≥97%
2-Chloro-4-(methylthio)pyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₆ClNOS
MW: 175.6359 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764127
CAS Number: 1463498-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR