1,2-Difluoro-4-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

71386-500MG 71386-1G 71386-5G 71386-25G
77333-600EA 161.71 USD
77333-600 77345-986 77369-612 77390-562
1,2-Difluoro-4-propoxybenzene ≥97%
1,2-Difluoro-4-propoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀F₂O
MW: 172.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27934423
CAS Number: 949027-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR