2-Bromo-1-ethoxy-3,4-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

71381-1G 71381-5G 71381-25G
77341-366EA 82.4 USD
77341-366 77369-610 77381-684
2-Bromo-1-ethoxy-3,4-difluorobenzene ≥97%
2-Bromo-1-ethoxy-3,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD30475317
CAS Number: 2092544-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR