2-Chloro-3,4-difluoro-5-iodo-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

71376-500MG 71376-1G 71376-5G 71376-25G
77333-598EA 280.16 USD
77333-598 77357-734 77369-606 77390-560
2-Chloro-3,4-difluoro-5-iodo-1-methoxybenzene ≥97%
2-Chloro-3,4-difluoro-5-iodo-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₄ClF₂IO
MW: 304.4603 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764109
CAS Number: 2901065-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR