(4-Chloro-2,3-difluoro-5-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

71374-500MG 71374-1G 71374-5G 71374-25G
77333-596EA 260.59 USD
77333-596 77357-732 77369-604 77390-558
(4-Chloro-2,3-difluoro-5-methoxyphenyl)(methyl)sulfane ≥97%
(4-Chloro-2,3-difluoro-5-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₇ClF₂OS
Storage Temperature: Ambient
MDL Number: MFCD34764108
CAS Number: 2901105-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR