1-Bromo-2,3-difluoro-5-methoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

71361-500MG 71361-1G 71361-5G
77336-566EA 388.31 USD
77336-566 77357-728 77381-678
1-Bromo-2,3-difluoro-5-methoxy-4-methylbenzene ≥97%
1-Bromo-2,3-difluoro-5-methoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34764100
CAS Number: 2901096-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR