3,6-Difluoro-2-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

70105-500MG 70105-1G 70105-5G
77339-620EA 603.58 USD
77339-620 77357-716 77381-668
3,6-Difluoro-2-formylbenzoic acid ≥97%
3,6-Difluoro-2-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₂O₃
MW: 186.112 g/mol
Storage Temperature: Ambient
CAS Number: 2138164-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR