2,5-Difluoro-3-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

70104-500MG 70104-1G 70104-5G 70104-25G
77333-586EA 182.31 USD
77333-586 77347-018 77369-600 77390-550
2,5-Difluoro-3-formylbenzoic acid ≥97%
2,5-Difluoro-3-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₂O₃
MW: 186.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764028
CAS Number: 2167760-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR