5-(tert-Butyl)-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69983-500MG 69983-1G 69983-5G 69983-25G
77333-584EA 263.68 USD
77333-584 77357-714 77369-598 77390-548
5-(tert-Butyl)-2-fluorobenzaldehyde ≥97%
5-(tert-Butyl)-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO
MW: 180.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24686599
CAS Number: 1123173-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR