2,3-Difluoro-5-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69682-500MG 69682-1G 69682-5G
77336-702EA 391.4 USD
77336-702 77357-710 77381-664
2,3-Difluoro-5-hydroxybenzaldehyde ≥97%
2,3-Difluoro-5-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28789018
CAS Number: 1806330-39-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR