1-(2-Ethoxy-5-methylphenyl)-2,2-difluoroethanone ≥97%

Supplier: AOBChem USA

Warning

69652-500MG 69652-1G 69652-5G 69652-25G
77333-576EA 244.11 USD
77333-576 77357-706 77369-590 77390-542
1-(2-Ethoxy-5-methylphenyl)-2,2-difluoroethanone ≥97%
1-(2-Ethoxy-5-methylphenyl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₂F₂O₂
MW: 214.209 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34764013
CAS Number: 1698589-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR