1-Ethoxy-2-iodo-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

69648-500MG 69648-1G 69648-5G 69648-25G
77333-574EA 143.17 USD
77333-574 77345-012 77369-588 77390-540
1-Ethoxy-2-iodo-4-methylbenzene ≥97%
1-Ethoxy-2-iodo-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO
MW: 262.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23728089
CAS Number: 1502858-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR