4-Ethyl-2-iodo-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

69635-500MG 69635-1G 69635-5G 69635-25G
77333-568EA 178.19 USD
77333-568 77346-622 77369-582 77390-534
4-Ethyl-2-iodo-1-methoxybenzene ≥97%
4-Ethyl-2-iodo-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO
Storage Temperature: Ambient
MDL Number: MFCD27979123
CAS Number: 868167-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR