(5-Ethyl-2-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

69634-500MG 69634-1G 69634-5G 69634-25G
77333-566EA 200.85 USD
77333-566 77357-702 77369-580 77390-532
(5-Ethyl-2-methoxyphenyl)(methyl)sulfane ≥97%
(5-Ethyl-2-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD22489267
CAS Number: 1822670-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR