5'-Fluoro-2,2''-dimethyl-1,1':3',1''-terphenyl ≥95%

Supplier: AOBChem USA

Warning

69623-500MG 69623-1G 69623-5G 69623-25G
77333-562EA 93.73 USD
77333-562 77342-488 77369-576 77381-660
5'-Fluoro-2,2''-dimethyl-1,1':3',1''-terphenyl ≥95%
5'-Fluoro-2,2''-dimethyl-1,1':3',1''-terphenyl

Precaución: for Research Use Only

Formula: C₂₀H₁₇F
MW: 276.347 g/mol
Storage Temperature: Ambient
CAS Number: 2807470-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR