5'-Fluoro-3,3''-dimethyl-1,1':3',1''-terphenyl ≥97%

Supplier: AOBChem USA

Warning

69622-500MG 69622-1G 69622-5G 69622-25G
77333-560EA 99.91 USD
77333-560 77342-884 77369-574 77381-658
5'-Fluoro-3,3''-dimethyl-1,1':3',1''-terphenyl ≥97%
5'-Fluoro-3,3''-dimethyl-1,1':3',1''-terphenyl

Precaución: for Research Use Only

Formula: C₂₀H₁₇F
MW: 276.347 g/mol
Storage Temperature: Ambient
CAS Number: 2807446-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR