5'-Ethyl-1,1':3',1''-terphenyl ≥97%

Supplier: AOBChem USA

Warning

69620-500MG 69620-1G 69620-5G 69620-25G
77333-558EA 97.85 USD
77333-558 77342-680 77369-570 77381-654
5'-Ethyl-1,1':3',1''-terphenyl ≥97%
5'-Ethyl-1,1':3',1''-terphenyl

Precaución: for Research Use Only

Formula: C₂₀H₁₈
Storage Temperature: Ambient
CAS Number: 10346-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR