5'-Fluoro-4,4''-bis(trifluoromethoxy)-1,1':3',1''-terphenyl ≥97%

Supplier: AOBChem USA

Warning

69619-500MG 69619-1G 69619-5G 69619-25G
77333-556EA 116.39 USD
77333-556 77343-462 77369-568 77381-652
5'-Fluoro-4,4''-bis(trifluoromethoxy)-1,1':3',1''-terphenyl ≥97%
5'-Fluoro-4,4''-bis(trifluoromethoxy)-1,1':3',1''-terphenyl

Precaución: for Research Use Only

Formula: C₂₀H₁₁F₇O₂
MW: 416.289 g/mol
Storage Temperature: Ambient
CAS Number: 2807463-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR