4,4'',5'-Trifluoro-1,1':3',1''-terphenyl ≥97%

Supplier: AOBChem USA

Warning

69616-500MG 69616-1G 69616-5G 69616-25G
77333-550EA 95.79 USD
77333-550 77342-630 77369-562 77381-646
4,4'',5'-Trifluoro-1,1':3',1''-terphenyl ≥97%
4,4'',5'-Trifluoro-1,1':3',1''-terphenyl

Precaución: for Research Use Only

Formula: C₁₈H₁₁F₃
Storage Temperature: Ambient
CAS Number: 2807442-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR