3-Ethoxy-4-fluoro-5-(methoxymethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69595-500MG 69595-1G 69595-5G
77340-374EA 973.35 USD
77340-374 77357-698 77381-642
3-Ethoxy-4-fluoro-5-(methoxymethyl)benzaldehyde ≥97%
3-Ethoxy-4-fluoro-5-(methoxymethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763997
CAS Number: 2901066-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR