4-Fluoro-3-methoxy-5-(methoxymethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

69587-500MG 69587-1G 69587-5G
77340-144EA 792.07 USD
77340-144 77357-694 77381-638
4-Fluoro-3-methoxy-5-(methoxymethyl)benzoic acid ≥97%
4-Fluoro-3-methoxy-5-(methoxymethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₄
MW: 214.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763989
CAS Number: 2901082-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR