2,2-Difluoro-1-(2-methyl-4-(trifluoromethyl)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

69567-500MG 69567-1G 69567-5G
77339-144EA 554.14 USD
77339-144 77357-690 77381-636
2,2-Difluoro-1-(2-methyl-4-(trifluoromethyl)phenyl)ethanone ≥97%
2,2-Difluoro-1-(2-methyl-4-(trifluoromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₇F₅O
Storage Temperature: Ambient
CAS Number: 1595788-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR