(2,4-Difluoro-3-hydroxyphenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

69536-500MG 69536-1G 69536-5G 69536-25G
77333-544EA 257.5 USD
77333-544 77357-688 77369-556 77390-528
(2,4-Difluoro-3-hydroxyphenyl)boronic acid ≥95%
(2,4-Difluoro-3-hydroxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28789159
CAS Number: 2894848-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR