1-Bromo-3-fluoro-4-iodo-2-(methoxymethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

69529-500MG 69529-1G 69529-5G 69529-25G
77333-542EA 141.11 USD
77333-542 77344-914 77369-554 77390-526
1-Bromo-3-fluoro-4-iodo-2-(methoxymethyl)benzene ≥97%
1-Bromo-3-fluoro-4-iodo-2-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₇BrFIO
MW: 344.947 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763976
CAS Number: 2807470-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR