3'-Bromo-2',6'-difluoro-[1,1'-biphenyl]-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

69517-500MG 69517-1G 69517-5G 69517-25G
77333-540EA 141.11 USD
77333-540 77344-912 77369-552 77390-524
3'-Bromo-2',6'-difluoro-[1,1'-biphenyl]-3-carbaldehyde ≥97%
3'-Bromo-2',6'-difluoro-[1,1'-biphenyl]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₇BrF₂O
MW: 297.095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32109679
CAS Number: 1976202-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR