(3'-Bromo-2',6'-difluoro-[1,1'-biphenyl]-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

69516-500MG 69516-1G 69516-5G 69516-25G
77333-538EA 161.71 USD
77333-538 77345-984 77369-550 77390-522
(3'-Bromo-2',6'-difluoro-[1,1'-biphenyl]-4-yl)methanol ≥97%
(3'-Bromo-2',6'-difluoro-[1,1'-biphenyl]-4-yl)methanol

Precaución: for Research Use Only

Formula: C₁₃H₉BrF₂O
MW: 299.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763971
CAS Number: 2901099-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR