3'-Bromo-2',6'-difluoro-[1,1'-biphenyl]-4-carbaldehyde ≥95%

Supplier: AOBChem USA

Warning

69515-500MG 69515-1G 69515-5G 69515-25G
77333-536EA 82.4 USD
77333-536 77342-164 77369-548 77381-632
3'-Bromo-2',6'-difluoro-[1,1'-biphenyl]-4-carbaldehyde ≥95%
3'-Bromo-2',6'-difluoro-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₇BrF₂O
MW: 297.095 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32109681
CAS Number: 1549716-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR