3-Bromo-2,4',6-trifluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

69511-500MG 69511-1G 69511-5G 69511-25G
77333-530EA 141.11 USD
77333-530 77344-908 77369-542 77390-516
3-Bromo-2,4',6-trifluoro-1,1'-biphenyl ≥97%
3-Bromo-2,4',6-trifluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₆BrF₃
Storage Temperature: Ambient
MDL Number: MFCD34763967
CAS Number: 2901099-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR