3-Bromo-2,6-difluoro-2'-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

69508-500MG 69508-1G 69508-5G 69508-25G
77333-528EA 136.99 USD
77333-528 77344-630 77369-540 77390-514
3-Bromo-2,6-difluoro-2'-methyl-1,1'-biphenyl ≥95%
3-Bromo-2,6-difluoro-2'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉BrF₂
MW: 283.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763964
CAS Number: 2901065-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR