3-Bromo-2,6-difluoro-4'-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

69502-500MG 69502-1G 69502-5G 69502-25G
77333-526EA 184.37 USD
77333-526 77347-330 77369-538 77390-512
3-Bromo-2,6-difluoro-4'-methyl-1,1'-biphenyl ≥95%
3-Bromo-2,6-difluoro-4'-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₉BrF₂
Storage Temperature: Ambient
MDL Number: MFCD34763958
CAS Number: 2901085-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR