3-Bromo-2,6-difluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

69499-500MG 69499-1G 69499-5G 69499-25G
77333-524EA 184.37 USD
77333-524 77347-328 77369-536 77390-510
3-Bromo-2,6-difluoro-1,1'-biphenyl ≥97%
3-Bromo-2,6-difluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇BrF₂
MW: 269.085 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763955
CAS Number: 2901105-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR