2,4-Difluoro-3-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

69489-500MG 69489-1G 69489-5G 69489-25G
77333-522EA 252.35 USD
77333-522 77357-684 77369-534 77390-508
2,4-Difluoro-3-(methylthio)benzoic acid ≥97%
2,4-Difluoro-3-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD24490933
CAS Number: 169310-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR