3-Bromo-5-fluoro-4-iodo-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

69462-500MG 69462-1G 69462-5G
77333-520EA 336.81 USD
77333-520 77357-682 77381-630
3-Bromo-5-fluoro-4-iodo-1,1'-biphenyl ≥97%
3-Bromo-5-fluoro-4-iodo-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇BrFI
MW: 376.991 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763946
CAS Number: 2901102-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR