4-Isobutoxy-2,6-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69416-500MG 69416-1G 69416-5G
77337-894EA 450.11 USD
77337-894 77357-676 77381-626
4-Isobutoxy-2,6-dimethylbenzaldehyde ≥97%
4-Isobutoxy-2,6-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD18426720
CAS Number: 1443312-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR