2-Iodo-1,3-dimethyl-5-propoxybenzene ≥97%

Supplier: AOBChem USA

Warning

69403-500MG 69403-1G 69403-5G 69403-25G
77333-518EA 116.39 USD
77333-518 77343-460 77369-532 77381-622
2-Iodo-1,3-dimethyl-5-propoxybenzene ≥97%
2-Iodo-1,3-dimethyl-5-propoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₅IO
MW: 290.141 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763928
CAS Number: 2901084-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR