(2,6-Dimethyl-4-propoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

69402-500MG 69402-1G 69402-5G 69402-25G
77333-516EA 243.08 USD
77333-516 77357-672 77369-530 77390-506
(2,6-Dimethyl-4-propoxyphenyl)(methyl)sulfane ≥97%
(2,6-Dimethyl-4-propoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₂H₁₈OS
Storage Temperature: Ambient
MDL Number: MFCD18427016
CAS Number: 1379322-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR