2-(Benzyloxy)-1-bromo-4-chloro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

69394-1G 69394-5G 69394-25G
77340-912EA 62.83 USD
77340-912 77369-528 77381-620
2-(Benzyloxy)-1-bromo-4-chloro-3-methylbenzene ≥97%
2-(Benzyloxy)-1-bromo-4-chloro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrClO
MW: 311.601 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763925
CAS Number: 2918879-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR