1-Bromo-4-chloro-2-isopropoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

69393-1G 69393-5G 69393-25G
77341-578EA 93.73 USD
77341-578 77369-526 77381-618
1-Bromo-4-chloro-2-isopropoxy-3-methylbenzene ≥97%
1-Bromo-4-chloro-2-isopropoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrClO
MW: 263.559 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763924
CAS Number: 2901085-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR