1-Bromo-4-chloro-2-ethoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

69392-1G 69392-5G 69392-25G
77341-288EA 81.37 USD
77341-288 77369-524 77381-616
1-Bromo-4-chloro-2-ethoxy-3-methylbenzene ≥97%
1-Bromo-4-chloro-2-ethoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763923
CAS Number: 2385847-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR