1-Chloro-2-fluoro-4-iodo-5-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

69374-500MG 69374-1G 69374-5G
77339-522EA 594.31 USD
77339-522 77357-668 77381-614
1-Chloro-2-fluoro-4-iodo-5-isopropoxybenzene ≥97%
1-Chloro-2-fluoro-4-iodo-5-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉ClFIO
MW: 314.523 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763920
CAS Number: 2807455-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR