4-Chloro-5-fluoro-2-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69372-500MG 69372-1G 69372-5G
77336-314EA 386.25 USD
77336-314 77357-666 77381-612
4-Chloro-5-fluoro-2-isopropoxybenzaldehyde ≥97%
4-Chloro-5-fluoro-2-isopropoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₂
MW: 216.637 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28627679
CAS Number: 1784255-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR