4-Chloro-5-fluoro-2-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

69371-500MG 69371-1G 69371-5G
77337-658EA 434.66 USD
77337-658 77357-664 77381-610
4-Chloro-5-fluoro-2-isopropoxybenzoic acid ≥97%
4-Chloro-5-fluoro-2-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₃
MW: 232.636 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763919
CAS Number: 1784176-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR