2-(Benzyloxy)-4-chloro-5-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

69364-500MG 69364-1G 69364-5G
77338-732EA 511.91 USD
77338-732 77357-656 77381-602
2-(Benzyloxy)-4-chloro-5-fluorobenzaldehyde ≥97%
2-(Benzyloxy)-4-chloro-5-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClFO₂
MW: 264.679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763915
CAS Number: 2807465-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR