2-(Benzyloxy)-4-chloro-5-fluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

69363-500MG 69363-1G 69363-5G
77339-520EA 594.31 USD
77339-520 77357-654 77381-600
2-(Benzyloxy)-4-chloro-5-fluorobenzoic acid ≥95%
2-(Benzyloxy)-4-chloro-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClFO₃
MW: 280.679 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34763914
CAS Number: 2159120-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR