(4-(Difluoromethoxy)phenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

69360-500MG 69360-1G 69360-5G 69360-25G
77333-512EA 161.71 USD
77333-512 77345-982 77369-520 77390-502
(4-(Difluoromethoxy)phenyl)(methyl)sulfane ≥97%
(4-(Difluoromethoxy)phenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈F₂OS
MW: 190.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31652671
CAS Number: 2177264-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR